Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQGILSPP AGLLSDDDVV VSPMFESTAA DLGSVVRKNL LSDCSVVSTS LEDKQQVPSE DSMEKVKVYL RVRPLLPSEL ERQEDQGCVR IENVETLVLQ APKDSFALKS NERGIGQATH RFTFSQIFGP EVGQASFFNL TVKEMVKDVL KGQNWLIYTY GVTNSGKTHT IQGTIKDGGI LPRSLALIFN SLQGQLHPTP DLKPLLSNEV IWLDSKQIRQ EEMKKLSLLN GGLQEEELST SLKRSVYIES RIGTSTSFDS GIAGLSSISQ CTSSSQLDET SHRWAQPDTA PLPVPANIRF SIWISFFEIY NELLYDLLEP PSQQRKRQTL RLCEDQNGNP YVKDLNWIHV QD (1-351 aa encoded by BC012999)
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GG10_2; Kinesin-like protein KIF20A; Mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); Rab6-interacting kinesin-like protein; Rabkinesin-6
Gene Aliases: KIF20A; MKLP2; RAB6KIFL
UniProt ID: (Human) O95235
Entrez Gene ID: (Human) 10112
Molecular Function: microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support