Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody specifically recognizes the endogenous KIF3A protein in mouse embryonic fibroblasts (MEFs) but not in KIF3A-/- lysate.
Immunogen sequence: MPINKSEKP ESCDNVKVVV RCRPLNEREK SMCYKQAVSV DEMRGTITVH KTDSSNEPPK TFTFDTVFGP ESKQLDVYNL TARPIIDSVL EGYNGTIFAY GQTGTGKTFT MEGVRAIPEL RGIIPNSFAH IFGHIAKAEG DTRFLVRVSY LGIYNEEVRD LLGKDQTQRL EVIERPDVGV YIKDLSAYVV NNADDMDRIM TLGHKNRSVG ATNMNEHSSR SHAIFTITIE CSEKGIDGNM HVRMGKLHLV DLAGSERQAK TGATGQRLKE ATKINLSLST LGNVISALVD GKSTHVPYRN SKLTRLLQDS LGGNSKTMMC ANIGPADYNY DETISTLRYA NRAKNIKNKA RINEDP (1-355 aa encoded by B C045542)
The kinesins constitute a large family of microtubule-dependent motor proteins which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. Members of the heterotrimeric kinesin II family of microtubule associated motors generally contain two different motor subunits from the KIF3 family, which includes KIF3A, B and C. KIF3 isoforms mediate anterograde transport of membrane bound organelles in neurons and melanosomes, transport between the endoplasmic reticulum and the Golgi, and transport of protein complexes within cilia and flagella required for their morphogenesis. KIF3A may influence neurogenesis at the level of embryonic cellular events, where the asymmetry of the genetic control circuit controlling left-right (L-R) axis determination is defined. Loss of KIF3A function in mice photoreceptors causes apoptotic cell death, suggesting that kinesin II mediated transport is required for proper cell fate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinesin family protein 3A; kinesin-II subunit; Kinesin-like protein KIF3A; Microtubule plus end-directed kinesin motor 3A; N-4 kinesin
Gene Aliases: FLA10; KIF3; KIF3A; Kifl; KLP-20; Kns3
UniProt ID: (Human) Q9Y496, (Mouse) P28741
Entrez Gene ID: (Human) 11127, (Mouse) 16568
Molecular Function: microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support