Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: ESVDALSEEL VQLRAQEKVH EMEKEHLNKV QTANEVKQAV EQQIQSHRET HQKQISSLRD EVEAKAKLIT DLQDQNQKMM LEQERLRVEH EKLKATDQEK SRKLHELTVM QDRREQARQD L
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: Conventional kinesin heavy chain; epididymis secretory protein Li 61; kif5b {ECO:0000312|EMBL:ABC25059.1}; kinesin 1 (110-120kD); kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; Kinesin-1 heavy chain; U-KHC; Ubiquitous kinesin heavy chain; UKHC
基因别名: AL022807; HEL-S-61; Khc; Khcs; KIF5B; KINH; KNS; KNS1; UKHC
UniProt ID: (Human) P33176, (Mouse) Q61768, (Rat) Q2PQA9
Entrez Gene ID: (Human) 3799, (Mouse) 16573, (Rat) 117550