Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody was raised against the C-terminal region of human KIFC1 protein, and recognizes 55-70 kDa of endogenous KIFC1 protein in multiple tissue lysates.
Immunogen sequence: GLLLFPSGP GGPSDPPTRL SLSRSDERRG TLSGAPAPPP RHDFSFDRVF PPGSGQDEVF EEIAMLVQSA LDGYPVCIFA YGQTGSGKTF TMEGGPGGDP QLEGLIPRAL RHLFSVAQEL SGQGWTYSFV ASYVEIYNET VRDLLATGTR KGQGGECEIR RAGPGSEELT VTNARYVPVS CEKEVDALLH LARQNRAVAR TAQNERSSRS HSVFQLQISG EHSSRGLQCG APLSLVDLAG SERLDPGLAL GPGERERLRE TQAINSSLST LGLVIMALSN KESHVPYRNS KLTYLLQNSL GGSAKMLMFV NISPLEENVS ESLNSLRFAS KVNQCVIGTA QANRK (382-725 aa encoded by BC000712)
Minus end-directed microtubule-dependent motor required for bipolar spindle formation (PubMed:15843429). May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kifc1; kifc1 {ECO:0000312|EMBL:AAH83827.1, ECO:0000312|RGD:1359118}; kinesin family c-terminal 5A; kinesin motor protein KIFC4; kinesin-like 2; kinesin-like 2a; Kinesin-like protein 2; Kinesin-like protein KIFC1; Kinesin-related protein 1; Kinesin-related protein HSET; kinesin-related protein KIFC5A; kinesin-related protein KRP1
Gene Aliases: HSET; 100042970; Gm4137; KIFC1; Kifc4; Kifc5a; Kifc5b; KNSL2; Knsl2a; Krp1; Tctex-7; Tctex-7A; Tctex7; Tctex7a
UniProt ID: (Human) Q9BW19, (Mouse) Q9QWT9, (Rat) Q5XI63
Entrez Gene ID: (Human) 3833, (Mouse) 100502766, (Rat) 294286
Molecular Function:
membrane trafficking regulatory protein
microtubule binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support