Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
KIR3DL1 are Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13. 4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several framework genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: 3DL1; AMB11; CD158 antigen-like family member E; CD158e; CD158e antigen; CD158E1/2; CD158E2; CL11; CL2; HLA-BW4-specific inhibitory NK cell receptor; Killer cell immunoglobulin-like receptor 3DL1; killer cell immunoglobulin-like receptor three domains long cytoplasmic tail 2; killer cell immunoglobulin-like receptor three domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1; killer Ig receptor; KIR antigen 3DL1; KIR G1; KIR3D; KIR3DS1; MGC119726; MGC119728; MGC126589; MGC126591; MHC class I NK cell receptor; natural killer cell receptor; Natural killer-associated transcript 3; NK receptor; NK-receptor; NKAT-3; NKAT10; p70; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clones CL-2/CL-11; p70 NK; p70 NK receptor CL-2/CL-11
Gene Aliases: CD158E; CD158E1; KIR; KIR3DL1; KIR3DL1/S1; KIR3DL2; NKAT-3; NKAT3; NKB1; NKB1B
UniProt ID: (Human) P43629
Entrez Gene ID: (Human) 3811
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support