Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in many species including rat, mouse.
Specificity of this antibody: Krueppel-like factor 2, 3, 4, 6, 7, 8, 12.
The Kruppel family of transcription factors (KLFs) are a set of C2H2-type zinc-finger DNA binding proteins that regulate gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AP-2 repressor; AP-2rep transcription factor; B-cell-derived protein 1; Basic krueppel-like factor; Basic krueppel-like factor 3; basic kruppel like factor; Basic Kruppel- like factor; basic Kruppel-like factor; basic kruppel-like factor 3; CACCC-box-binding protein BKLF; core promoter element binding protein; Core promoter element-binding protein; endothelial Kruppel-like zinc finger protein; Epithelial zinc finger protein EZF; GC-rich binding factor; GC-rich sites-binding factor GBF; gut Kruppel-like factor; Gut-enriched krueppel-like factor; immediate early response, erythropoietin 1; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Krueppel-like factor 2; Krueppel-like factor 3; Krueppel-like factor 4; Krueppel-like factor 6; Krueppel-like factor 7; Krueppel-like factor 8; Kruppel-like factor 12; Kruppel-like factor 2 (lung); Kruppel-like factor 3 (basic); Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); Kruppel-like factor 6; Kruppel-like factor 7 (ubiquitous); Kruppel-like factor 8; Kruppel-like factor 8, pseudogene 1; Kruppel-like factor LKLF; Kruppel-like zinc finger protein Zf9; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor; Proto-oncogene BCD1; protooncogene B-cell derived 1; suppression of tumorigenicity 12 (prostate); Suppressor of tumorigenicity 12 protein; TEF-2; transcript ch138; Transcription factor Zf9; Transcriptional repressor AP-2rep; Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; Zinc finger protein 741
Gene Aliases: 2700063E05Rik; 9830124P08Rik; 9930027G08Rik; A830097P10Rik; Aa1017; AI225908; AI315654; AI448727; AL024007; AP-2rep; AP2REP; B130052C06Rik; BCD1; BKLF; BKLF3; C86813; CBA1; COPEB; CPBP; D530033K05Rik; EZF; FM2; FM6; GBF; GKLF; HSPC122; Ierepo1; Ierepo3; KLF12; KLF2; KLF3; KLF4; KLF6; KLF7; KLF8; Klf8-ps1; LKLF; PAC1; R75280; RGD1565831; ST12; Tef-2; UKLF; ZF9; Zie; ZNF74; ZNF741
UniProt ID: (Human) Q9Y4X4, (Human) Q9Y5W3, (Human) P57682, (Human) O43474, (Human) Q99612, (Human) O75840, (Human) O95600, (Mouse) O35738, (Mouse) Q60843, (Rat) Q9ET58, (Mouse) Q60980, (Mouse) Q60793, (Mouse) Q99JB0, (Mouse) Q8BLM0
Entrez Gene ID: (Human) 11278, (Human) 10365, (Human) 51274, (Human) 9314, (Human) 1316, (Human) 8609, (Human) 11279, (Mouse) 16597, (Rat) 306110, (Mouse) 16598, (Rat) 306330, (Mouse) 16599, (Rat) 114845, (Mouse) 16600, (Rat) 114505, (Mouse) 23849, (Rat) 58954, (Mouse) 93691, (Rat) 363243, (Mouse) 245671, (Rat) 302582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support