Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Transcription regulator of erythrocyte development that probably serves as a general switch factor during erythropoiesis. Is a dual regulator of fetal-to-adult globin switching. Binds to the CACCC box in the beta-globin gene promoter and acts as a preferential activator of this gene. Furthermore, it binds to the BCL11A promoter and activates expression of BCL11A, which in turn represses the HBG1 and HBG2 genes. This dual activity ensures that, in most adults, fetal hemoglobin levels are low. Able to activate CD44 and AQP1 promoters. When sumoylated, acts as a transcriptional repressor by promoting interaction with CDH2/MI2beta and also represses megakaryocytic differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EKLF; endothelial Kruppel-like zinc finger protein; Epithelial zinc finger protein EZF; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; gut Kruppel-like factor; Gut-enriched krueppel-like factor; Krueppel-like factor 1; Krueppel-like factor 2; Krueppel-like factor 4; Kruppel-like factor 1 (erythroid); Kruppel-like factor 2 (lung); Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); Kruppel-like factor LKLF; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor; monoclonal antibody A3D8; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1
Gene Aliases: CDAN4; EKLF; EZF; GKLF; HBFQTL6; INLU; KLF1; KLF2; KLF4; LKLF; Nan; Zie
UniProt ID: (Human) Q13351, (Human) Q9Y5W3, (Human) O43474, (Mouse) Q60843, (Rat) Q9ET58, (Mouse) Q60793
Entrez Gene ID: (Human) 10661, (Human) 10365, (Human) 9314, (Rat) 304666, (Mouse) 16596, (Mouse) 16598, (Rat) 306330, (Mouse) 16600, (Rat) 114505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support