Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody is stable for 24 months.
Positive Control: HeLa or 293T cells.Human lymph node, spleen or colon. Cellular Localization: Nucleus.
Specificity Comments: Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
Predicted to react with Mouse and Rat
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AP-2 repressor; AP-2rep transcription factor; BHLHB24; ID; KLF12 zinc finger transcriptional repressor; Krueppel-like factor 12; Kruppel-like factor 12; Transcriptional repressor AP-2rep
Gene Aliases: AP-2rep; AP2REP; HSPC122; KLF12
UniProt ID: (Human) Q9Y4X4
Entrez Gene ID: (Human) 11278
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support