Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QMLPPQDAHD LGMPPAESQS LLVLGSQDSL VSQPDSQEGP FLPEQPGPAP QTVEKNSRPQ EGTGRRGSSE ARPYCCNYEN CGKAYTKRSH LVSHQRKHTG ERPYSCNWES CSWSFFRSDE LRRHMRVHTR YRPYKCDQCS REFMRSDHLK QHQKTHRPGP SDPQANNNNG EQDSPPAAGP; Positive Samples: U-87MG, HepG2, MCF7, Mouse brain; Cellular Location: Nucleus
KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5'-CACCC-3' sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ40160; germ cell specific zinc finger protein; Germ cell-specific zinc finger protein; Krueppel-like factor 17; Kruppel-like factor 17; novel zinc-finger protein; Zinc finger protein 393; ZLF393
Gene Aliases: 7420700M05Rik; AA420409; AU043468; C85123; D4Ertd561e; Gzf; KLF17; Zfp393; ZNF393
UniProt ID: (Human) Q5JT82, (Mouse) Q8CFA7
Entrez Gene ID: (Human) 128209, (Mouse) 75753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support