Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (intestinal Kruppel-like factor; basic transcription element binding protein 2; Basic transcription element-binding protein 2; BTE-binding protein 2; Colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5C isoform; Krueppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; Transcription factor BTEB2
Gene Aliases: 4930520J07Rik; BTEB2; CKLF; IKLF; KLF5
UniProt ID: (Human) Q13887, (Mouse) Q9Z0Z7
Entrez Gene ID: (Human) 688, (Mouse) 12224
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support