Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATRVLSMSA RLGPVPQPPA PQDEPVFAQL KPVLGAANPA RDAALFPGEE LKHAHHRPQA QPAPAQAPQP AQPPATGPRL PPEDLVQTRC EMEKYLTPQL PPVPIIPEHK KYRRDSASVV DQFFTDTEGL PYSINMNVFL PDITHLRTGL YKSQRPCVTH IKTEPVAIFS HQSETTAPPP APTQALPEFT SIFSSHQTAA PEVNNIFIKQ ELPTPDLHLS VPTQQGHLYQ LLNTPDLDMP SSTNQTAAMD TLNVSMSAAM AGLNTHTSAV PQTAVKQFQG MPPCTYTMPS QFLPQQATYF PPSPPSSEPG SPDRQAEMLQ NLTPPPSYAA TIASKLAIHN PNLPTTLPVN
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (intestinal Kruppel-like factor; basic transcription element binding protein 2; Basic transcription element-binding protein 2; BTE-binding protein 2; Colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5C isoform; Krueppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; Transcription factor BTEB2
Gene Aliases: 4930520J07Rik; BTEB2; CKLF; IKLF; KLF5
UniProt ID: (Human) Q13887, (Mouse) Q9Z0Z7
Entrez Gene ID: (Human) 688, (Mouse) 12224
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support