Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EWTQLKPNNL SCDLPEERYR HEIAHDGQRI YILGGGTSWT AYSLNKIHAY NLETNAWEEI ATKPHEKIGF PAARRCHSCV QIK
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kelch domain-containing protein 10; PNAS-119; scruin like at the midline homolog
Gene Aliases: KIAA0265; KLHDC10; slim
UniProt ID: (Human) Q6PID8
Entrez Gene ID: (Human) 23008
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support