Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like 10; kelch-like family member 10; Kelch-like protein 10; testicular tissue protein Li 104
Gene Aliases: 4921517C11Rik; KLHL10; SPGF11
UniProt ID: (Human) Q6JEL2, (Rat) Q6JEL3, (Mouse) Q9D5V2
Entrez Gene ID: (Human) 317719, (Rat) 303533, (Mouse) 66720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support