Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGIMAPKDI MTNTHAKSIL NSMNSLRKSN TLCDVTLRVE QKDFPAHRIV LAACSDYFCA MFTSELSEKG KPYVDIQGLT ASTMEILLDF VYTETVHVTV ENVQELLPAA CLLQLKGVKQ ACCEFLESQL DPSNCLGIRD FAETHNCVDL MQAAEVFSQK HFPEVVQHEE FILLSQGEVE KLIKCDEIQV DSEEPVFEAV INWVKHAKKE REESLPNLLQ YVRMPLLTPR YITDVIDAEP FIRCSLQCRD LVDEAKKFHL RPELRSQMQG PRTRARLGAN EVLLVVGGFG SQQSPIDVVE KYDPKTQEWS FLPSITRKRR YVASVSLHDR IYVIGGYDGR SRLSSVECLD YTADEDGVWY SVAPMNVRRG LAGATTLGDM IYVSGGFDGS RRHTSMERYD PNIDQWSMLG DMQTAREGAG LVVASGVIYC LGGYDGLNIL NSVEKYDPHT GHWTNVTPMA TKRSGAGVAL LNDHIYVVGG FDGTAHLSSV EAYNIRTDSW TTVTSMTTPR CYVGATVLRG RLYAIAGYDG NSLLSSIECY DPIIDSWEVV TSMGTQRCDA GVCVLREK
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CUL3-interacting protein 1; DKIR homolog; FLJ27152; hDKIR; kelch-like family member 12; Kelch-like protein 12; kelch-like protein C3IP1
Gene Aliases: C3IP1; DKIR; KLHL12
UniProt ID: (Human) Q53G59
Entrez Gene ID: (Human) 59349
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support