Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actinfilin; kelch-like 17; kelch-like family member 17; Kelch-like protein 17; RP11-54O7.6
Gene Aliases: AF; KLHL17
UniProt ID: (Human) Q6TDP4
Entrez Gene ID: (Human) 339451
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support