Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: RTDKENRVKN LSEVFDCIRF RLMTEKYFKD HVEKDDIIKS NPDLQKKIKV LKDAFAGKLP EPSKNAAKTG AGEVNGDVGD EDLLPGYLND IPRHGMFVKD
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kel-like protein 23; kelch repeat and BTB (POZ) domain containing 10; Kelch repeat and BTB domain-containing protein 10; kelch-like family member 41; Kelch-like protein 41; Kelch-related protein 1; sarcomeric muscle protein; Sarcosin
Gene Aliases: KBTBD10; KLHL41; KRP1; SARCOSIN
UniProt ID: (Human) O60662
Entrez Gene ID: (Human) 10324
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support