Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
KLRB1 (Killer cell lectin like receptor subfamily B, member 1, CD-161) protein is classified as a type II membrane protein because it has an external C terminus. KLRB1 cell surface antigen is expressed by almost all NK cells and in a small subset of CD3+ve T cells. KLRB1 is a homodimeric cell surface protein, comprising two chains with molecular weights ranging from 40-44kDa. KLRB1 plays an inhibitory role on natural killer (NK) cells. Activation of KLRB1 leads to acid sphingomyelnase/SMPD1 stimulation and an increase in intracellular ceramide. Moreover, there is also an activation of AKT1/PKB and RPS6KA1/RSK1 kinase stimulation, and T cell proliferation by anti-CD3. KLRB1 acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope and to the N-acetyllactosamine epitope. KLRB1 also binds to CLEC2D/LLT1 as a ligand, and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen 3.2.3; CD161 antigen-like family member A; CD161 antigen-like family member B; CD161a; CD161b; Immunoreceptor NKR-P1E; Killer cell lectin-like receptor subfamily B member 1; Killer cell lectin-like receptor subfamily B member 1A; killer cell lectin-like receptor subfamily B member 1B; Killer cell lectin-like receptor subfamily B member 1B allele A; killer cell lectin-like receptor subfamily B member 1B allele C; killer cell lectin-like receptor subfamily B member 1F; Killer cell lectin-like receptor subfamily B member 1G; klrb1b {ECO:0000312|EMBL:AAQ11375.1}; klrb1b {ECO:0000312|RGD:2975}; MGC138614; natural killer cell receptor; Natural killer cell surface protein NKR-P1B allele RNK/SD/BN/F344; natural killer cell surface protein NKR-P1B allele TO; Natural killer cell surface protein NKR-P1G; Natural killer cell surface protein P1-3.2.3; Natural killer lectin-like receptor 1E; natural killer lectin-like receptor protein 1 e; NKR-P1 3.2.3; NKR-P1A
Gene Aliases: Klrb1; Klrb1a; Klrb1b; Klrb1d; Klrb1f; Klrb1g; Klrb6; NKR-P1A; Nkr-p1b; Nkr-p1e; NKR-P1G; Nkrp-1e; Nkrp1a; Nkrp1b; Nkrp1e; Nkrp1g
UniProt ID: (Rat) Q0ZUP0, (Rat) P27471, (Rat) A4KWA1
Entrez Gene ID: (Rat) 689817, (Rat) 362443, (Rat) 25192
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support