Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • KLRG1 Antibodies

            Invitrogen

            KLRG1 Monoclonal Antibody (2F1), Pacific Orange™, eBioscience™

            18 Published Figures
            18 References
            View all (61) KLRG1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 79-5893-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            738,00
            100 µg
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite KLRG1 Monoclonal Antibody (2F1), Pacific Orange™, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (18)
            Application
            KLRG1 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 19

            KLRG1 Antibody (79-5893-82) in Flow

            C57BL/6 mouse splenocytes were stained with NK1.1 Monoclonal Antibody, APC (Product # 17-5941-82) and 0.25 µg of Syrian Hamster IgG Isotype Control, Pacific Orange (Product # 79-4914-81) (left) or 0.25 µg of KLRG1 Monoclonal Antibody, Pacific Orange (right). Cells in the lymphocyte gate were used for analysis.
            View Product

            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)
            KLRG1 Antibody in Flow Cytometry (Flow)

            Product Details

            79-5893-82

            Applications
            Tested Dilution
            Publications

            Flow Cytometry (Flow)

            0.5 µg/test
            View 17 publications 17 publications

            Functional Assay (Functional)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Mouse

            Host/Isotype

            Syrian hamster / IgG

            Class

            Monoclonal

            Type

            Antibody

            Clone

            2F1

            Conjugate

            Pacific Orange™
            • Alexa Fluor 488 
            • Alexa Fluor 532 
            • Alexa Fluor 561
            • Alexa Fluor 647 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 711
            • eFluor 450
            • eFluor 506
            • FITC
            • Functional Grade
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation

            Excitation/Emission Max

            400/551 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2815359

            Product Specific Information

            Description: This 2F1 monoclonal antibody reacts with the mouse Killer cell Lectin-like Receptor G1 (KLRG1), also known as Mast cell Function-associated Antigen (MAFA). KLRG1 is a homodimer of glycosylated 30-38 kDa subunits and contains a cytoplasmic motif similar to the immunoreceptor tyrosine-based inhibitory motif (ITIM). Rat MAFA was identified as an antigen specific to rat mast cells; however, the expression of mouse KLRG1/MAFA using 2F1 has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. This antigen is expressed on approximately one-third of mouse NK cells and a subset of T cells. MHC class I molecules regulate KLRG1 via interactions with class I-specific inhibitory Ly49 molecules and SHP-1 signaling. Although KLRG1 and Ly49 are both lectin-like inhibitory receptors that are regulated by class I MHC expression, the effects of this on cell surface expression of these molecules are opposing, and the underlying regulatory mechanisms distinct.

            Applications Reported: This 2F1 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This 2F1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Pacific Orange emits at 551 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome.

            Excitation: 405 nm; Emission: 551 nm; Laser: Violet Laser

            Target Information

            KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              How to use the Panel Builder

              Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

              Bioinformatics

              Protein Aliases: Killer cell lectin-like receptor subfamily G member 1; Mast cell function-associated antigen 2F1; natural killer cell receptor

              View more View less

              Gene Aliases: 2F1-Ag; Klrg1; MAFA; MAFA-L

              View more View less

              UniProt ID: (Mouse) O88713

              View more View less

              Entrez Gene ID: (Mouse) 50928

              View more View less

              Function(s)

              carbohydrate binding transmembrane signal receptor

              Process(es)

              immune system process cell surface receptor signaling pathway innate immune response
              It has to be done as per old AB suggested Products section.
              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              • Price & Freight Policy
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • German Supply Chain Due Diligence Act
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Legal Notice
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              germany | deutschland flag icon
              Germany | Deutschland

              Your items have has been added!


              Host server : magellan-search-green-56756dfb8b-nsvbw:80/100.66.79.248:80.
              git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.31.1-2025.06.25-1.0