Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLNCDVLSHF PNLLSHPKEK INKEAVWFLS NITAGNQQQV QAVIDAGLIP MIIHQLAKGD FGTQKEAAWA ISNLTISGRK DQVEYLVQQN VIPPFCNLLS VKDSQVVQVV LDGLKNILIM AGDEASTIAE II
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: importin alpha 4; Importin alpha Q2; importin alpha-3; importin subunit alpha-3; Importin subunit alpha-4; importin-alpha-Q2; karyopherin (importin) alpha 3; karyopherin alpha 3 (importin alpha 4); Karyopherin subunit alpha-3; predicted role in nuclear import; Qip2; RP11-432M24.3; SRP1-gamma
Gene Aliases: hSRP1; importin; IPOA4; KPNA3; QIP2; SRP1; SRP1gamma; SRP4
UniProt ID: (Human) O00505, (Mouse) O35344
Entrez Gene ID: (Human) 3839, (Mouse) 16648, (Rat) 361055
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support