Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDAMASPGKD NYRMKSYKNK ALNPQEMRRR REEEGIQLRK QKREEQLFKR RNVYLPRNDE SMLESPIQDP DISSTVPIPE EEVVTTDMVQ MIFSNNADQQ LTATQKFRKL LSKEPNPPID QVIQKPGVVQ RFVKFLERNE NCTLQFEAAW ALTNIASGTF LHTKVVIETG AVPIFIKLLN SEHEDVQEQA VWALGNIAGD NAECRDFVLN CEILPPLLEL LTNSNRLTTT RNAVWALSNL CRGKNPPPNF SKVSPCLNVL SRLLFSSDPD VLADVCWALS YLSDGPNDKI QAVIDSGVCR RLVELLMHND YKVVSPALRA VGNIVTGDDI QTQVILNCSA LPCLLHLLSS PKESIRKEAC WTVSNITAGN RAQIQAVIDA NIFPVLIEIL QKAEFRTRKE AAWAITNATS GGTPEQIRYL VALGCIKPLC DLLTVMDSKI VQVALNGLEN ILRLGEQESK QNGIGINPYC ALIEEAYGLD KIEFLQSHEN QEIYQKAFDL IEHYFGVEED DPSIVPQVDE NQQQFIFQQQ EAPMDGFQL
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC) which consists of 60-100 proteins and is probably 120 million daltons in molecular size. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion; larger molecules are transported by an active process. Most nuclear proteins contain short basic amino acid sequences known as nuclear localization signals (NLSs). KPNA5 protein belongs to the importin alpha protein family and is thought to be involved in NLS-dependent protein import into the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: importin alpha 6; Importin subunit alpha-6; karyopherin alpha 5 (importin alpha 6); Karyopherin subunit alpha-5
Gene Aliases: IPOA6; KPNA5; SRP6
UniProt ID: (Human) O15131
Entrez Gene ID: (Human) 3841
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support