Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MAMSQESLTF KDVFVDFTLE EWQQLDSAQK NLYRDVMLEN YSHLVSVGYL VAKPDVIFRL GPGEESWMAD GGTPVRTCAG EDRPDVSIFA SCILKCCY
This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KRAB box domain-containing protein 4; KRAB domain-containing protein 4; putative zinc finger transcription factor; zinc finger family member 673; zinc finger protein 673
Gene Aliases: KRBOX4; ZNF673
UniProt ID: (Human) Q5JUW0
Entrez Gene ID: (Human) 55634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support