Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat small intestine tissue lysate.
PA5-34520 can be used with blocking peptide PEP-1563.
KREMEN1 is a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. This protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dickkopf receptor; Kremen protein 1; Kringle domain-containing transmembrane protein 1; kringle-coding gene marking the eye and the nose; Kringle-containing protein marking the eye and the nose
Gene Aliases: AV002070; KREMEN; KREMEN1; KRM1
UniProt ID: (Human) Q96MU8, (Mouse) Q99N43, (Rat) Q924S4
Entrez Gene ID: (Human) 83999, (Mouse) 84035, (Rat) 114107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support