Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The Keratin multigene family is made of "soft" epithelial cytokeratins and "hard" hair Keratins. While the epithelial cytokeratins are involved in the layering and formation of epithelia, the hair Keratins are responsible for creating nails and hair. There are two types of Keratins: the acidic class I Keratin proteins and the basic/neutral class II Keratin proteins. Keratin 84, also known as KRT84, HB4 or KRTHB4, is a 600 amino acid protein that is expressed in hair follicles. Keratin 84 is a basic/neutral type II Keratin protein which heterodimerizes with type I Keratins to create hair and nails. The gene encoding Keratin 84 maps to human chromosome 12 which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis and Kniest dysplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hard keratin, type II, 4; K84; keratin 84, type II; keratin, hair, basic, 4; Keratin, type II cuticular Hb4; Keratin-84; type II hair keratin 4; Type II hair keratin Hb4; type II keratin Kb24; Type-II keratin Kb24
Gene Aliases: HB4; Kb24; KRT84; KRTHB4
UniProt ID: (Human) Q9NSB2
Entrez Gene ID: (Human) 3890, (Rat) 315320
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support