Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PAPPLPPSAT PPSPLHPSPQ CTRQQKNFNL PASHYYKYKQ QFIFPDVVPV PETPTRAPQV ILHPVTSNPI LEGNPLLQIE VEPTSENEEV
KSR2 serine/threonine kinase or kinase suppressor of Ras 2 specifically regulates the activity of MEKK3 and COT but not other members of the MAP3K family. KSR interacts with a regulatory Raf molecule in cis to induce a conformational switch of MEK, facilitating MEK's phosphorylation by a separate catalytic Raf molecule in trans. Blocks MAP3K8 kinase activity and MAP3K8- mediated signaling. Acts as a negative regulator of MAP3K3- mediated activation of ERK, JNK and NF-kappa-B pathways, inhibiting MAP3K3-mediated interleukin-8 production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hKSR2; Kinase suppressor of Ras 2
Gene Aliases: KSR2
UniProt ID: (Human) Q6VAB6
Entrez Gene ID: (Human) 283455
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support