Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: SAQGRQKES GSKWKVFIDQ INRSLENYEP CSSQNCSCYH GVIEEDLTPF RGGISRKMMA EVVRRKLGTH YQITKNRLYR ENDCMFPSRC SGVEHFILEV IGRLPDMEMV INVRDYPQVP KWMEPAIPVF SFSKTSEYHD IMYPAWTFWE GGPAVWPIYP TGLGRWDLFR EDLVRSAAQW PWKKKNSTAY FRGSRTSPER DPLILLSRKN PKLVDAEYTK NQAWKSMKDT LGKPAAKDVH LVDHCKYKYL FNFRGVAASF RFKHLFLCGS LVFHVGDEWL EFFYPQLKPW VHYIPVKTDL SNVQELLQFV KANDDVAQEI AERGSQFIRN HLQMDDITCY WENLLSEYSK FLSYNVTRRK GYDQIIPKML KTEL (20-392 aa encoded by BC048810)
KTELC1 also termed as CLP46 is a glucosyltransferase, which implicates in Protein modification and glycosylation. KTELC1 encodes a 392 amino acids protein and its molecular weight is 46 kDa, but is exists glycosylation modification. KTELC1 is localized in endoplasmic reticulum lumen and abundantly expressed in liver, brain, heart, skeletal muscle, spleen, kidney, placenta, lung and peripheral blood leukocyte. KTELC1 may regulation in cell proliferation and Notch signal pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 9630046K23Rik; CAP10-like 46 kDa protein; CAP10-like protein; hCLP46; hRumi; KDELC family like 1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; x 010 protein
Gene Aliases: C3orf9; CLP46; hCLP46; KDELCL1; KTELC1; MDS010; MDSRP; POGLUT1; Rumi; UNQ490/PRO1006
UniProt ID: (Human) Q8NBL1
Entrez Gene ID: (Human) 56983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support