Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments, and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif. The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain potassium channel protein-1; IA; MBK1; megencephaly; MGC126782; MGC138385; MKI; potassium (K+) channel protein voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 1; potassium voltage gated channel shaker related subfamily member 1; potassium voltage gated channel, shaker related subfamily, member 1; Potassium voltage-gated channel subfamily A member 1; potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia); RBKI; RCK1; Voltage-gated K(+) channel HuKI; Voltage-gated potassium channel HBK1; Voltage-gated potassium channel subunit Kv1.1
Gene Aliases: AEMK; AI840627; EA1; HBK1; HUK1; Kca1-1; Kcna; KCNA1; Kcpvd; KV1.1; MBK1; mceph; Mk-1; MK1; RBK1; Shak
UniProt ID: (Human) Q09470, (Mouse) P16388, (Rat) P10499
Entrez Gene ID: (Human) 3736, (Mouse) 16485, (Rat) 24520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support