Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitution: 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. The gene is brain-specific, and located in the neocortex and the striatum. It may be involved in cellular excitability of restricted neurons in the central nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEC2; Brain-specific eag-like channel 2; ELK channel 1; ether-a-go-go K(+) channel family member; Ether-a-go-go-like potassium channel 1; potassium channel, voltage gated eag related subfamily H, member 4; Potassium voltage-gated channel subfamily H member 4; potassium voltage-gated channel, subfamily H (eag-related), member 4; rElk1; Voltage-gated delayed rectifier potassium channel KCNH4; Voltage-gated potassium channel subunit Kv12.3
Gene Aliases: BEC2; ELK1; Elk3; KCNH4; Kv12.3
UniProt ID: (Human) Q9UQ05, (Rat) Q9R1T9
Entrez Gene ID: (Human) 23415, (Rat) 114032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support