Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide sequence: YAERIGADPD DILGSNHTYF KNIPIGFWWA VVTMTTLGYG DMYPKTWSGM
Sequence homology: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KSHIIID; potassium channel, voltage gated Shaw related subfamily C, member 3; Potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; Shaw-related voltage-gated potassium channel protein 3; Voltage-gated potassium channel KCNC3; voltage-gated potassium channel protein KV3.3; Voltage-gated potassium channel subunit Kv3.3
Gene Aliases: KCNC3; KSHIIID; KV3.3; SCA13
UniProt ID: (Human) Q14003
Entrez Gene ID: (Human) 3748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support