Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NTLDRYPDTL LGSSEKEFFY DADSGEYFFD RDPDMFRHVL NFYRTGRLHC PRQECIQAFD EELAFYGLVP ELVGDCCLEE YRDRKKENAE RLAEDEEAEQ
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha-subunits (KV alpha), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV beta subunits that modify the gating properties of the coexpressed KV alpha subunits. Differences exist in the patterns of trafficking, biosynthetic processing and surface expression of the major KV1 subunits (KV1. 1, KV1. 2 and KV1. 4) expressed in rat and human brain, suggesting that the individual protein subunits are highly regulated to control for the assembly and formation of functional neuronal channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: potassium channel, voltage gated Shal related subfamily D, member 1; Potassium voltage-gated channel subfamily D member 1; potassium voltage-gated channel, Shal-related subfamily, member 1; Shal-type potassium channel; Voltage-gated potassium channel subunit Kv4.1
Gene Aliases: KCND1; KV4.1
UniProt ID: (Human) Q9NSA2
Entrez Gene ID: (Human) 3750
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support