Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
MA5-31783 has been tested in indirect ELISA.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes-shaker, shaw, shab, and shal-have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential. This member mediates a rapidly inactivating, A-type outward potassium current which is not under the control of the N terminus as it is in Shaker channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-type voltage-gated potassium channel KCND2; potassium channel, voltage gated Shal related subfamily D, member 2; Potassium voltage-gated channel subfamily D member 2; Voltage-gated potassium channel subunit Kv4.2; voltage-sensitive potassium channel
Gene Aliases: KCND2; KIAA1044; KV4.2; RK5
UniProt ID: (Human) Q9NZV8
Entrez Gene ID: (Human) 3751
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support