Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide Sequence: FRRIRTLKGK LARQHPEAFS HIPEASFLEE EDEDPIPPST TTTIATSEQS TGSCDTSPDT VSPSLSPGFE DLSHVQAGSP AINGRSQTDD EEMTGE
The function of the C19orf50 protein has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KxDL motif-containing protein 1; UPF0459 protein C19orf50
Gene Aliases: BORCS4; C19orf50; KXD1; KXDL; MST096; MSTP096
UniProt ID: (Human) Q9BQD3
Entrez Gene ID: (Human) 79036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support