Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPSSLELPA DTVQRIAAEL KCHPTDERVA LHLDEEDKLR HFRECFYIPK IQDLPPVDLS LVNKDENAIY FLGNSLGLQP KMVKTYLEEE LDKWAKIAAY GHEVGKRPWI TGDESIVGLM KDIVGANEKE IALMNALTVN LHLLMLSFFK PTPKRYKILL EAKAFPSDHY AIESQLQLHG LNIEESMRMI KPREGEETLR IEDILEVIEK EGDSIAVILF SGVHFYTGQH FNIPAITKAG QAKGCYVGFD LAHAVGNVEL YLHDWGVDFA CWCSYKYLNA GAGGIAGAFI HEKHAHTIKP ARSEFFN
Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KYNU; Kynureninase; L-kynurenine hydrolase
Gene Aliases: KYNU; KYNUU
UniProt ID: (Human) Q16719
Entrez Gene ID: (Human) 8942
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support