Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: LLPQNDTRLD PEAYGSPCAR GSQPWQVSLF NGLSFHCAGV LVDQSWVLTA
Sequence homology: Human: 100%
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: breast normal epithelial cell associated serine protease; kallikrein 10 protein 1; kallikrein 10 protein 12; kallikrein 10 protein 13; kallikrein 10 protein 2; kallikrein 10 protein 3; kallikrein 10 protein 4; kallikrein 10 protein 5; kallikrein 10 protein 7; kallikrein 10 protein 8; kallikrein 10 protein 9; Kallikrein-10; kallikrein-related peptidase 10; Normal epithelial cell-specific 1; Protease serine-like 1; protease, serine-like, 1
Gene Aliases: KLK10; NES1; PRSSL1
UniProt ID: (Human) O43240
Entrez Gene ID: (Human) 5655
Molecular Function: protease serine protease protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support