Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; Kallikrein related-peptidase 14; Kallikrein-14; Kallikrein-like protein 6; kallikrein-related peptidase 14; KLK-14; KLK-L6; mGK14
Gene Aliases: GK14; KLK-L6; KLK14; KLKL6
UniProt ID: (Human) Q9P0G3, (Mouse) Q8CGR5
Entrez Gene ID: (Human) 43847, (Rat) 308562, (Mouse) 317653
Molecular Function: protease serine protease protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support