Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MWDLVLSIA LSVGCTGAVP LIQSRIVGGW ECEKHSQPWQ VAVYSHGWAH CGGVLVHPQW VLTAAHCLKK NSQVWLGRHN LFEPEDTGQR VPVSHSFPHP LYNMSLLKHQ SLRPDEDSSH DLMLLRLSEP AKITDVVKVL GLPTQEPALG TTCYASGWGS IEPEEFLRPR SLQCVSLHLL SNDMCARAYS EKVTEFMLCA GLWTGGKDTC GGDSGGPLVC NGVLQGITSW GPEPCALPEK PAVYTKVVHY RKWIKDTIAA NP (1-261 aa encoded by BC005196)
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glandular kallikrein 2; Glandular kallikrein-1; hGK-1; kallikrein 2, prostatic; Kallikrein-2; kallikrein-related peptidase 2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2
Gene Aliases: hGK-1; hK2; KLK2; KLK2A2
UniProt ID: (Human) P20151
Entrez Gene ID: (Human) 3817
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support