Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
FIGURE: 1 / 4
Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HK5; kallikrein 5; Kallikrein L2; Kallikrein like 2; Kallikrein-5; Kallikrein-like protein 2; kallikrein-related peptidase 5; Kallikrein5; KLK 5; KLK L2; KLK-5; KLK-L2; KLKL 2; Stratum corneum tryptic enzyme
Gene Aliases: KLK-L2; KLK5; KLKL2; SCTE; UNQ570/PRO1132
UniProt ID: (Human) Q9Y337
Entrez Gene ID: (Human) 25818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support