Product References
Poly-L-ornithine blocks the inhibitory effects of fibronectin on oligodendrocyte differentiation and promotes myelin repair.
Neural regeneration research
Xiong YJ,Soomro SH,Huang ZH,Yu PP,Ping J,Fu H
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Sat Apr 01 00:00:00 UTC 2023
Transformation of primary murine peritoneal mast cells by constitutive KIT activation is accompanied by loss of Cdkn2a/Arf expression.
Frontiers in immunology
Capellmann S,Sonntag R,Schüler H,Meurer SK,Gan L,Kauffmann M,Horn K,Königs-Werner H,Weiskirchen R,Liedtke C,Huber M
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Wed Apr 19 00:00:00 UTC 2023
B-cell antigen receptor expression and phosphatidylinositol 3-kinase signaling regulate genesis and maintenance of mouse chronic lymphocytic leukemia.
Haematologica
Schmid VK,Khadour A,Ahmed N,Brandl C,Nitschke L,Rajewsky K,Jumaa H,Hobeika E
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Inhibition of LncRNA Vof-16 expression promotes nerve regeneration and functional recovery after spinal cord injury.
Neural regeneration research
Zhang XM,Zeng LN,Yang WY,Ding L,Chen KZ,Fu WJ,Zeng SQ,Liang YR,Chen GH,Wu HF
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Sat Jan 01 00:00:00 UTC 2022
Application of an instructive hydrogel accelerates re-epithelialization of xenografted human skin wounds.
Scientific reports
Sparks HD,Mandla S,Vizely K,Rosin N,Radisic M,Biernaskie J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Sat Aug 20 00:00:00 UTC 2022
Application of an instructive hydrogel accelerates re-epithelialization of xenografted human skin wounds.
Scientific reports
Sparks HD,Mandla S,Vizely K,Rosin N,Radisic M,Biernaskie J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Sat Aug 20 00:00:00 UTC 2022
Visualization of individual cell division history in complex tissues using iCOUNT.
Cell stem cell
Denoth-Lippuner A,Jaeger BN,Liang T,Royall LN,Chie SE,Buthey K,Machado D,Korobeynyk VI,Kruse M,Munz CM,Gerbaulet A,Simons BD,Jessberger S
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Nov 04 00:00:00 UTC 2021
Epicardial placement of human MSC-loaded fibrin sealant films for heart failure: Preclinical efficacy and mechanistic data.
Molecular therapy : the journal of the American Society of Gene Therapy
Fields L,Ito T,Kobayashi K,Ichihara Y,Podaru MN,Hussain M,Yamashita K,Machado V,Lewis-McDougall F,Suzuki K
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Wed Aug 04 00:00:00 UTC 2021
Environmental oxygen regulates astrocyte proliferation to guide angiogenesis during retinal development.
Development (Cambridge, England)
Perelli RM,O'Sullivan ML,Zarnick S,Kay JN
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Sat May 01 00:00:00 UTC 2021
HDAC1 Regulates Neuronal Differentiation.
Frontiers in molecular neuroscience
Nieto-Estevez V,Changarathil G,Adeyeye AO,Coppin MO,Kassim RS,Zhu J,Hsieh J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry (PFA fixed)
Tue Feb 01 00:00:00 UTC 2022
Macrophage LAMTOR1 Deficiency Prevents Dietary Obesity and Insulin Resistance Through Inflammation-Induced Energy Expenditure.
Frontiers in cell and developmental biology
Ying L,Zhang M,Ma X,Si Y,Li X,Su J,Yin J,Bao Y
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
Transcription factor old astrocyte specifically induced substance is a novel regulator of kidney fibrosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Yamamoto A,Morioki H,Nakae T,Miyake Y,Harada T,Noda S,Mitsuoka S,Matsumoto K,Tomimatsu M,Kanemoto S,Tanaka S,Maeda M,Conway SJ,Imaizumi K,Fujio Y,Obana M
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
HOXA11 plays critical roles in disease progression and response to cytarabine in AML.
Oncology reports
Fu JF,Shih LY,Yen TH
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
Thrombospondin-2 spatiotemporal expression in skeletal fractures.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Zondervan RL,Jenkins DC,Reicha JD,Hankenson KD
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri Jan 01 00:00:00 UTC 2021
MicroRNA-384 inhibits nasopharyngeal carcinoma growth and metastasis via binding to Smad5 and suppressing the Wnt/β-catenin axis.
Cytotechnology
Zeng X,Liao H,Wang F
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Apr 01 00:00:00 UTC 2021
Requirement of DNMT1 to orchestrate epigenomic reprogramming for NPM-ALK-driven lymphomagenesis.
Life science alliance
Redl E,Sheibani-Tezerji R,Cardona CJ,Hamminger P,Timelthaler G,Hassler MR,Zrimšek M,Lagger S,Dillinger T,Hofbauer L,Draganić K,Tiefenbacher A,Kothmayer M,Dietz CH,Ramsahoye BH,Kenner L,Bock C,Seiser C,Ellmeier W,Schweikert G,Egger G
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
A booster dose enhances immunogenicity of the COVID-19 vaccine candidate ChAdOx1 nCoV-19 in aged mice.
Med (New York, N.Y.)
Silva-Cayetano A,Foster WS,Innocentin S,Belij-Rammerstorfer S,Spencer AJ,Burton OT,Fra-Bidó S,Le Lee J,Thakur N,Conceicao C,Wright D,Barrett J,Evans-Bailey N,Noble C,Bailey D,Liston A,Gilbert SC,Lambe T,Linterman MA
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Mar 12 00:00:00 UTC 2021
Age-related neurodegeneration and cognitive impairments of NRMT1 knockout mice are preceded by misregulation of RB and abnormal neural stem cell development.
Cell death & disease
Catlin JP,Marziali LN,Rein B,Yan Z,Feltri ML,Schaner Tooley CE
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Oct 28 00:00:00 UTC 2021
Ten-eleven translocation protein 1 modulates medulloblastoma progression.
Genome biology
Kim H,Kang Y,Li Y,Chen L,Lin L,Johnson ND,Zhu D,Robinson MH,McSwain L,Barwick BG,Yuan X,Liao X,Zhao J,Zhang Z,Shu Q,Chen J,Allen EG,Kenney AM,Castellino RC,Van Meir EG,Conneely KN,Vertino PM,Jin P,Li J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Apr 29 00:00:00 UTC 2021
Mutant ASXL1 induces age-related expansion of phenotypic hematopoietic stem cells through activation of Akt/mTOR pathway.
Nature communications
Fujino T,Goyama S,Sugiura Y,Inoue D,Asada S,Yamasaki S,Matsumoto A,Yamaguchi K,Isobe Y,Tsuchiya A,Shikata S,Sato N,Morinaga H,Fukuyama T,Tanaka Y,Fukushima T,Takeda R,Yamamoto K,Honda H,Nishimura EK,Furukawa Y,Shibata T,Abdel-Wahab O,Suematsu M,Kitamura T
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Tue Mar 23 00:00:00 UTC 2021
An mTORC1-dependent switch orchestrates the transition between mouse spermatogonial stem cells and clones of progenitor spermatogonia.
Cell reports
Suzuki S,McCarrey JR,Hermann BP
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Tue Feb 16 00:00:00 UTC 2021
An mTORC1-dependent switch orchestrates the transition between mouse spermatogonial stem cells and clones of progenitor spermatogonia.
Cell reports
Suzuki S,McCarrey JR,Hermann BP
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Tue Feb 16 00:00:00 UTC 2021
IL-1beta promotes the age-associated decline of beta cell function.
iScience
Böni-Schnetzler M,Méreau H,Rachid L,Wiedemann SJ,Schulze F,Trimigliozzi K,Meier DT,Donath MY
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Nov 19 00:00:00 UTC 2021
CDCP1 promotes compensatory renal growth by integrating Src and Met signaling.
Life science alliance
Kajiwara K,Yamano S,Aoki K,Okuzaki D,Matsumoto K,Okada M
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry (PFA fixed)
Thu Apr 01 00:00:00 UTC 2021
Targeting Phosphatidylserine Enhances the Anti-tumor Response to Tumor-Directed Radiation Therapy in a Preclinical Model of Melanoma.
Cell reports
Budhu S,Giese R,Gupta A,Fitzgerald K,Zappasodi R,Schad S,Hirschhorn D,Campesato LF,De Henau O,Gigoux M,Liu C,Mazo G,Deng L,Barker CA,Wolchok JD,Merghoub T
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
CXCR4 Regulates Temporal Differentiation via PRC1 Complex in Organogenesis of Epithelial Glands.
International journal of molecular sciences
Kim J,Lee SW,Park K
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Sun Jan 10 00:00:00 UTC 2021
Delta-like 4 is required for pulmonary vascular arborization and alveolarization in the developing lung.
JCI insight
Xia S,Menden HL,Townley N,Mabry SM,Johnston J,Nyp MF,Heruth DP,Korfhagen T,Sampath V
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Apr 08 00:00:00 UTC 2021
Adult neural stem cells have latent inflammatory potential that is kept suppressed by Tcf4 to facilitate adult neurogenesis.
Science advances
Shariq M,Sahasrabuddhe V,Krishna S,Radha S,Bellampalli R,Dwivedi A,Cheramangalam R,Reizis B,Hébert J,Ghosh HS
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Sat May 01 00:00:00 UTC 2021
Notch Signaling between Cerebellar Granule Cell Progenitors.
eNeuro
Adachi T,Miyashita S,Yamashita M,Shimoda M,Okonechnikov K,Chavez L,Kool M,Pfister SM,Inoue T,Kawauchi D,Hoshino M
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Jul 02 00:00:00 UTC 2021
Developmental Role of Adenosine Kinase in the Cerebellum.
eNeuro
Gebril H,Wahba A,Zhou X,Lai T,Alharfoush E,DiCicco-Bloom E,Boison D
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Jul 02 00:00:00 UTC 2021
Shikonin Derivatives from Onsoma visianii Decrease Expression of Phosphorylated STAT3 in Leukemia Cells and Exert Antitumor Activity.
Nutrients
Todorovic Z,Milovanovic J,Arsenijevic D,Vukovic N,Vukic M,Arsenijevic A,Djurdjevic P,Milovanovic M,Arsenijevic N
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Wed Mar 31 00:00:00 UTC 2021
Field size effects on DNA damage and proliferation in normal human cell populations irradiated with X-ray microbeams.
Scientific reports
Ojima M,Ito A,Usami N,Ohara M,Suzuki K,Kai M
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri Mar 26 00:00:00 UTC 2021
Dissecting phenotypic transitions in metastatic disease via photoconversion-based isolation.
eLife
Sela Y,Li J,Kuri P,Merrell AJ,Li N,Lengner C,Rompolas P,Stanger BZ
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Feb 23 00:00:00 UTC 2021
NEIL3-deficiency increases gut permeability and contributes to a pro-atherogenic metabolic phenotype.
Scientific reports
Karlsen TR,Kong XY,Holm S,Quiles-Jiménez A,Dahl TB,Yang K,Sagen EL,Skarpengland T,S Øgaard JD,Holm K,Vestad B,Olsen MB,Aukrust P,Bjørås M,Hov JR,Halvorsen B,Gregersen I
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Tue Oct 05 00:00:00 UTC 2021
GTSE1 is possibly involved in the DNA damage repair and cisplatin resistance in osteosarcoma.
Journal of orthopaedic surgery and research
Xie C,Xiang W,Shen H,Shen J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Dec 07 00:00:00 UTC 2021
TWIK-1 BAC-GFP Transgenic Mice, an Animal Model for TWIK-1 Expression.
Cells
Kwon O,Yang H,Kim SC,Kim J,Sim J,Lee J,Hwang EM,Shim S,Park JY
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Oct 14 00:00:00 UTC 2021
Targeting Feedforward Loops Formed by Nuclear Receptor RORγ and Kinase PBK in mCRPC with Hyperactive AR Signaling.
Cancers
Zhang X,Huang Z,Wang J,Ma Z,Yang J,Corey E,Evans CP,Yu AM,Chen HW
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Apr 01 00:00:00 UTC 2021
Gene Signatures Detect Damaged Liver Sinusoidal Endothelial Cells in Chronic Liver Diseases.
Frontiers in medicine
Verhulst S,van Os EA,De Smet V,Eysackers N,Mannaerts I,van Grunsven LA
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Wed Nov 10 00:00:00 UTC 2021
Imbalanced Activation of Wnt-/β-Catenin-Signaling in Liver Endothelium Alters Normal Sinusoidal Differentiation.
Frontiers in physiology
Koch PS,Sandorski K,Heil J,Schmid CD,Kürschner SW,Hoffmann J,Winkler M,Staniczek T,de la Torre C,Sticht C,Schledzewski K,Taketo MM,Trogisch FA,Heineke J,Géraud C,Goerdt S,Olsavszky V
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Oct 22 00:00:00 UTC 2021
An in vivo model allowing continuous observation of human vascular formation in the same animal over time.
Scientific reports
Tsukada Y,Muramatsu F,Hayashi Y,Inagaki C,Su H,Iba T,Kidoya H,Takakura N
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Jan 12 00:00:00 UTC 2021
Human Lung Stem Cell-Based Alveolospheres Provide Insights into SARS-CoV-2-Mediated Interferon Responses and Pneumocyte Dysfunction.
Cell stem cell
Katsura H,Sontake V,Tata A,Kobayashi Y,Edwards CE,Heaton BE,Konkimalla A,Asakura T,Mikami Y,Fritch EJ,Lee PJ,Heaton NS,Boucher RC,Randell SH,Baric RS,Tata PR
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Dec 03 00:00:00 UTC 2020
Egr2-guided histone H2B monoubiquitination is required for peripheral nervous system myelination.
Nucleic acids research
Wüst HM,Wegener A,Fröb F,Hartwig AC,Wegwitz F,Kari V,Schimmel M,Tamm ER,Johnsen SA,Wegner M,Sock E
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri Sep 18 00:00:00 UTC 2020
Genetic manipulation of LKB1 elicits lethal metastatic prostate cancer.
The Journal of experimental medicine
Hermanova I,Zúñiga-García P,Caro-Maldonado A,Fernandez-Ruiz S,Salvador F,Martín-Martín N,Zabala-Letona A,Nuñez-Olle M,Torrano V,Camacho L,Lizcano JM,Talamillo A,Carreira S,Gurel B,Cortazar AR,Guiu M,López JI,Martinez-Romero A,Astobiza I,Valcarcel-Jimenez L,Lorente M,Arruabarrena-Aristorena A,Velasco G,Gomez-Muñoz A,Suárez-Cabrera C,Lodewijk I,Flores JM,Sutherland JD,Barrio R,de Bono JS,Paramio JM,Trka J,Graupera M,Gomis RR,Carracedo A
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Mon Jun 01 00:00:00 UTC 2020
Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Developmental cell
Vila Ellis L,Cain MP,Hutchison V,Flodby P,Crandall ED,Borok Z,Zhou B,Ostrin EJ,Wythe JD,Chen J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Mon Mar 09 00:00:00 UTC 2020
Multimodal single-cell analysis reveals distinct radioresistant stem-like and progenitor cell populations in murine glioma.
Glia
Alexander J,LaPlant QC,Pattwell SS,Szulzewsky F,Cimino PJ,Caruso FP,Pugliese P,Chen Z,Chardon F,Hill AJ,Spurrell C,Ahrendsen D,Pietras A,Starita LM,Hambardzumyan D,Iavarone A,Shendure J,Holland EC
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Dec 01 00:00:00 UTC 2020
FGF2 modulates simultaneously the mode, the rate of division and the growth fraction in cultures of radial glia.
Development (Cambridge, England)
Ledesma-Terrón M,Peralta-Cañadas N,Míguez DG
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri Jul 24 00:00:00 UTC 2020
NPC1 deficiency impairs cerebellar postnatal development of microglia and climbing fiber refinement in a mouse model of Niemann-Pick disease type C.
Development (Cambridge, England)
Boyle BR,Melli SE,Altreche RS,Padron ZM,Yousufzai FAK,Kim S,Vasquez MD,Carone DM,Carone BR,Soto I
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry (Frozen)
Mon Aug 03 00:00:00 UTC 2020
Novel enhanced GFP-positive congenic inbred strain establishment and application of tumor-bearing nude mouse model.
Cancer science
Lan Q,Chen Y,Dai C,Li S,Fei X,Dong J,Shen Y,Dai X,Lu Z,Liu B,Wang Q,Wang H,Zhou Z,Ji X,Wang Z,Huang Q
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Oct 01 00:00:00 UTC 2020
Mycobacterium tuberculosis-infected alveolar epithelial cells modulate dendritic cell function through the HIF-1α-NOS2 axis.
Journal of leukocyte biology
Rodrigues TS,Alvarez ARP,Gembre AF,Forni MFPAD,de Melo BMS,Alves Filho JCF,Câmara NOS,Bonato VLD
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Radiation Induced Metabolic Alterations Associate With Tumor Aggressiveness and Poor Outcome in Glioblastoma.
Frontiers in oncology
Gupta K,Vuckovic I,Zhang S,Xiong Y,Carlson BL,Jacobs J,Olson I,Petterson XM,Macura SI,Sarkaria J,Burns TC
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Mon Sep 28 00:00:00 UTC 2020
Radiation Induced Metabolic Alterations Associate With Tumor Aggressiveness and Poor Outcome in Glioblastoma.
Frontiers in oncology
Gupta K,Vuckovic I,Zhang S,Xiong Y,Carlson BL,Jacobs J,Olson I,Petterson XM,Macura SI,Sarkaria J,Burns TC
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Mon Sep 28 00:00:00 UTC 2020
Agrin Promotes Limbal Stem Cell Proliferation and Corneal Wound Healing Through Hippo-Yap Signaling Pathway.
Investigative ophthalmology & visual science
Hou L,Fu W,Liu Y,Wang Q,Wang L,Huang Y
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Mon May 11 00:00:00 UTC 2020
New Insights into the Pathogenesis of Diabetic Nephropathy: Proximal Renal Tubules Are Primary Target of Oxidative Stress in Diabetic Kidney.
Acta histochemica et cytochemica
Haraguchi R,Kohara Y,Matsubayashi K,Kitazawa R,Kitazawa S
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Apr 28 00:00:00 UTC 2020
A single-cell transcriptomic and anatomic atlas of mouse dorsal raphe Pet1 neurons.
eLife
Okaty BW,Sturrock N,Escobedo Lozoya Y,Chang Y,Senft RA,Lyon KA,Alekseyenko OV,Dymecki SM
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Mon Jun 22 00:00:00 UTC 2020
Elevated Serum Amino Acids Induce a Subpopulation of Alpha Cells to Initiate Pancreatic Neuroendocrine Tumor Formation.
Cell reports. Medicine
Smith DK,Kates L,Durinck S,Patel N,Stawiski EW,Kljavin N,Foreman O,Sipos B,Solloway MJ,Allan BB,Peterson AS
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Tue Aug 25 00:00:00 UTC 2020
Metastasis-initiating cells induce and exploit a fibroblast niche to fuel malignant colonization of the lungs.
Nature communications
Pein M,Insua-Rodríguez J,Hongu T,Riedel A,Meier J,Wiedmann L,Decker K,Essers MAG,Sinn HP,Spaich S,Sütterlin M,Schneeweiss A,Trumpp A,Oskarsson T
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Mar 20 00:00:00 UTC 2020
Aripiprazole reduces liver cell division.
PloS one
Pirc Marolt T,Kramar B,Bulc Rozman K,Šuput D,Milisav I
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Tue Dec 08 00:00:00 UTC 2020
Regeneration of the pulmonary vascular endothelium after viral pneumonia requires COUP-TF2.
Science advances
Zhao G,Weiner AI,Neupauer KM,de Mello Costa MF,Palashikar G,Adams-Tzivelekidis S,Mangalmurti NS,Vaughan AE
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Sun Nov 01 00:00:00 UTC 2020
Transcriptome analysis of basic fibroblast growth factor treated stem cells isolated from human exfoliated deciduous teeth.
Heliyon
Nowwarote N,Manokawinchoke J,Kanjana K,Fournier BPJ,Sukarawan W,Osathanon T
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Mon Jun 01 00:00:00 UTC 2020
Comparative Analysis of Age-Related Changes in Lacrimal Glands and Meibomian Glands of a C57BL/6 Male Mouse Model.
International journal of molecular sciences
Yoon CH,Ryu JS,Hwang HS,Kim MK
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu Jun 11 00:00:00 UTC 2020
The effects of hedgehog ligand neutralising antibody 5E1 in a mouse model of endometriosis.
BMC research notes
Cousins FL,Farley JK,Kerrigan R,Mukherjee S,Darzi S,Gargett CE,Deane JA
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri Sep 25 00:00:00 UTC 2020
CD103+ cDC1 and endogenous CD8+ T cells are necessary for improved CD40L-overexpressing CAR T cell antitumor function.
Nature communications
Kuhn NF,Lopez AV,Li X,Cai W,Daniyan AF,Brentjens RJ
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Wed Dec 02 00:00:00 UTC 2020
B Cell Activating Factor (BAFF) Is Required for the Development of Intra-Renal Tertiary Lymphoid Organs in Experimental Kidney Transplantation in Rats.
International journal of molecular sciences
Steines L,Poth H,Herrmann M,Schuster A,Banas B,Bergler T
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Wed Oct 28 00:00:00 UTC 2020
A discrete subtype of neural progenitor crucial for cortical folding in the gyrencephalic mammalian brain.
eLife
Matsumoto N,Tanaka S,Horiike T,Shinmyo Y,Kawasaki H
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Tue Apr 21 00:00:00 UTC 2020
A discrete subtype of neural progenitor crucial for cortical folding in the gyrencephalic mammalian brain.
eLife
Matsumoto N,Tanaka S,Horiike T,Shinmyo Y,Kawasaki H
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Tue Apr 21 00:00:00 UTC 2020
Treatment of mice with a ligand binding blocking anti-CD28 monoclonal antibody improves healing after myocardial infarction.
PloS one
Gladow N,Hollmann C,Ramos G,Frantz S,Kerkau T,Beyersdorf N,Hofmann U
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Mon Jul 06 00:00:00 UTC 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Thu Dec 17 00:00:00 UTC 2020
Discrete populations of isotype-switched memory B lymphocytes are maintained in murine spleen and bone marrow.
Nature communications
Riedel R,Addo R,Ferreira-Gomes M,Heinz GA,Heinrich F,Kummer J,Greiff V,Schulz D,Klaeden C,Cornelis R,Menzel U,Kröger S,Stervbo U,Köhler R,Haftmann C,Kühnel S,Lehmann K,Maschmeyer P,McGrath M,Naundorf S,Hahne S,Sercan-Alp Ö,Siracusa F,Stefanowski J,Weber M,Westendorf K,Zimmermann J,Hauser AE,Reddy ST,Durek P,Chang HD,Mashreghi MF,Radbruch A
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Fri May 22 00:00:00 UTC 2020
Changes in Liver Mechanical Properties and Water Diffusivity During Normal Pregnancy Are Driven by Cellular Hypertrophy.
Frontiers in physiology
Garczyńska K,Tzschätzsch H,Kühl AA,Morr AS,Lilaj L,Häckel A,Schellenberger E,Berndt N,Holzhütter HG,Braun J,Sack I,Guo J
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Fri Dec 18 00:00:00 UTC 2020
Interleukin-13 disrupts type 2 pneumocyte stem cell activity.
JCI insight
Glisinski KM,Schlobohm AJ,Paramore SV,Birukova A,Moseley MA,Foster MW,Barkauskas CE
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Jan 16 00:00:00 UTC 2020
Interleukin-13 disrupts type 2 pneumocyte stem cell activity.
JCI insight
Glisinski KM,Schlobohm AJ,Paramore SV,Birukova A,Moseley MA,Foster MW,Barkauskas CE
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Thu Jan 16 00:00:00 UTC 2020
Microbiota-Sourced Purines Support Wound Healing and Mucous Barrier Function.
iScience
Lee JS,Wang RX,Goldberg MS,Clifford GP,Kao DJ,Colgan SP
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry (Paraffin)
Fri Jun 26 00:00:00 UTC 2020
Eradication of tumor growth by delivering novel photothermal selenium-coated tellurium nanoheterojunctions.
Science advances
Chen S,Xing C,Huang D,Zhou C,Ding B,Guo Z,Peng Z,Wang D,Zhu X,Liu S,Cai Z,Wu J,Zhao J,Wu Z,Zhang Y,Wei C,Yan Q,Wang H,Fan D,Liu L,Zhang H,Cao Y
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Wed Apr 01 00:00:00 UTC 2020
Deficiency in the secreted protein Semaphorin3d causes abnormal parathyroid development in mice.
The Journal of biological chemistry
Singh A,Mia MM,Cibi DM,Arya AK,Bhadada SK,Singh MK
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunocytochemistry
Fri May 24 00:00:00 UTC 2019
Myoepithelial Cells of Submucosal Glands Can Function as Reserve Stem Cells to Regenerate Airways after Injury.
Cell stem cell
Tata A,Kobayashi Y,Chow RD,Tran J,Desai A,Massri AJ,McCord TJ,Gunn MD,Tata PR
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Immunohistochemistry
Thu May 03 00:00:00 UTC 2018
De Novo Epigenetic Programs Inhibit PD-1 Blockade-Mediated T Cell Rejuvenation.
Cell
Ghoneim HE,Fan Y,Moustaki A,Abdelsamed HA,Dash P,Dogra P,Carter R,Awad W,Neale G,Thomas PG,Youngblood B
Published figure using Ki-67 monoclonal antibody (Product # 53-5698-82) in Flow Cytometry
Thu Jun 29 00:00:00 UTC 2017