Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Several different potassium channels are known to be involved with electrical signaling in the nervous system. One class is activated by depolarization whereas a second class is not. The latter are referred to as inwardly rectifying K+ channels, and they have a greater tendency to allow potassium to flow into the cell rather than out of it. This asymmetry in potassium ion conductance plays a key role in the excitability of muscle cells and neurons. The protein encoded by this gene is an integral membrane protein and member of the inward rectifier potassium channel family. The encoded protein has a small unitary conductance compared to other members of this protein family. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BIR11; Brain inwardly rectifying K(+) channel 2; Hippocampal inward rectifier; hippocampal inward rectifier potassium channel; HIR; HIRK2; HRK1; Inward rectifier K(+) channel Kir2.3; Inward rectifier potassium channel 4; IRK-3; IRK3; Kir 2.3; Potassium channel, inwardly rectifying subfamily J member 4; potassium channel, inwardly rectifying subfamily J, member 4; potassium inwardly-rectifying channel, subfamily J, member 4
Gene Aliases: HIR; HIRK2; HRK1; IRK-3; IRK3; Kcnf2; KCNJ4; Kir2.3; MB-IRK3
UniProt ID: (Human) P48050, (Rat) P52190
Entrez Gene ID: (Human) 3761, (Mouse) 16520, (Rat) 116649
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support