Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-activated inward rectifier potassium channel 3; G protein-coupled inward rectifier potassium channel; GIRK-3; Inward rectifier K(+) channel Kir3.3; inwardly rectifier K(+) channel Kir3.3; inwardly rectifier K+ channel KIR3.3; Potassium channel, inwardly rectifying subfamily J member 9; potassium channel, inwardly rectifying subfamily J, member 9; potassium inwardly-rectifying channel, subfamily J, member 9
Gene Aliases: 1700085N21Rik; GIRK3; KCNJ9; KIR3.3; mbGIRK3
UniProt ID: (Human) Q92806, (Rat) Q63511, (Mouse) P48543
Entrez Gene ID: (Human) 3765, (Rat) 116560, (Mouse) 16524
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support