Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total KCNJ15 protein.
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-sensitive inward rectifier potassium channel 15; Inward rectifier K(+) channel Kir1.3; Inward rectifier K(+) channel Kir4.2; inward rectifier K+ channel KIR4.2; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15
Gene Aliases: 4930414N08Rik; AI182284; AI267127; IRKK; KCNJ14; KCNJ15; KIR1.3; KIR4.2
UniProt ID: (Human) Q99712, (Mouse) O88932
Entrez Gene ID: (Human) 3772, (Mouse) 16516
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support