Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: VRAQPMPHSI LSTQTSRC
Kirrel3 Short Isoform 3 is a variant of the Kirrel3 protein, which is a member of the immunoglobulin superfamily involved in cell adhesion and signaling. This specific isoform, as a shorter version, may have distinct functional or regulatory roles compared to the full-length protein, particularly in the development and maintenance of neural connections. Mutations or dysregulation of Kirrel3 have been associated with neurodevelopmental disorders, including intellectual disability and autism spectrum disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kin of IRRE-like protein 3; Kin of irregular chiasm-like protein 3; membrane protein mKirre; mKirre; Nephrin-like protein 2; X kin of IRRE like 3
Gene Aliases: 1500010O20Rik; 2900036G11Rik; Kiaa1867; Kirrel3; mKIAA1867; NEPH2; SST4
UniProt ID: (Mouse) Q8BR86
Entrez Gene ID: (Mouse) 67703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support