Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Does not react with Mouse, Rat, chicken
The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 70 kDa subunit of Ku antigen; 86 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC75; CTC85; CTCBF; DNA repair protein XRCC5; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen, 70kDa; Ku autoantigen, 80kDa; Ku70; Ku80; Ku86; Ku86 autoantigen related protein 1; Lupus Ku autoantigen protein p70; Lupus Ku autoantigen protein p86; Nuclear factor IV; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 5; X-ray repair cross-complementing protein 6
Gene Aliases: CTC75; CTCBF; G22P1; G22P2; KARP-1; KARP1; KU70; KU80; Ku86; KUB2; ML8; NFIV; TLAA; XRCC5; XRCC6
UniProt ID: (Human) P13010, (Human) P12956
Entrez Gene ID: (Human) 7520, (Human) 2547
Molecular Function:
DNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support