Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: QYLQVTSCPK IPSSPDLKKS RSASTISKSD YMEIQEGVNN SNEDFREENL KTANCTLANT NYVNITKMLT DV
Kcna2 is a voltage-gated potassium channel (KV) that belongs to the 6-TM family of potassium channel and also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). The Kcna2 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. Kcna2 contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. Further, Kcna2 belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. The coding region of the Kcna2 gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1. Diseases associated with KCNA2 include Epileptic Encephalopathy (Early Infantile, 32) and Undetermined Early-Onset Epileptic Encephalopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MK2; NGK1; Potassium (K+) channel protein alpha 2, voltage dependent; potassium channel, voltage gated shaker related subfamily A, member 2; potassium voltage gated channel shaker related subfamily member 2; Potassium voltage-gated channel subfamily A member 2; potassium voltage-gated channel, shaker-related subfamily, member 2; RAK; RBK2; RCK5; RP11-284N8.1; Voltage-gated K(+) channel HuKIV; Voltage-gated potassium channel HBK5; voltage-gated potassium channel protein Kv1.2; Voltage-gated potassium channel subunit Kv1.2
Gene Aliases: Akr6a4; BK2; EIEE32; ENSMUSG00000074335; Gm10672; HBK5; HK4; HUKIV; Kca1-2; KCNA2; KV1.2; Mk-2; MK2; NGK1; RBK2
UniProt ID: (Human) P16389, (Mouse) P63141, (Rat) P63142
Entrez Gene ID: (Human) 3737, (Mouse) 16490, (Rat) 25468
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support