Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KQT like 2; KQT-like 2; KVEBN1; Neuroblastoma-specific potassium channel subunit alpha KvLQT2; Potassium channel subunit alpha KvLQT2; potassium channel, voltage gated KQT-like subfamily Q, member 2; Potassium voltage-gated channel subfamily KQT member 2; potassium voltage-gated channel, KQT-like subfamily, member 2; potassium voltage-gated channel, subfamily Q, member 2; Voltage-gated potassium channel subunit Kv7.2
Gene Aliases: BFNC; EBN; EBN1; ENB1; HNSPC; KCNA11; KCNQ2; KQT2; KV7.2; Nmf134
UniProt ID: (Human) O43526, (Rat) O88943, (Mouse) Q9Z351
Entrez Gene ID: (Human) 3785, (Rat) 170848, (Mouse) 16536
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support