Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
ABS 005-16-02 detects L-Ficolin from human samples.
ABS 005-16-02 has been successfully used in ELISA and Western blot procedures.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin B; Ficolin-2; Ficolin-B; Ficolin-beta; Hucolin; L-ficolin; RP11-263F14.2; Serum lectin p35
Gene Aliases: EBP-37; FCN2; FCNL; ficolin-2; P35
UniProt ID: (Human) Q15485
Entrez Gene ID: (Human) 2220
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support