Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PVGWCKEHR RTLITPPGYP NVKHFSWDKY LEETNSLPAP ARAFKVKPPH GFQKKMKLEV VDKRNPMFIR VATVADTDDH RVKVHFDGWN NCYDYWIDAD SPDIHPVGWC SKTGHPLQPP LSPLELMEAS EHGGCSTPGC KGIGHFKRAR HLGPHSAANC PYSEINLNKD RIFPDRLSGE MPPASPSFPR NKRTDANESS SSPEIRDQHA DDVKEDFEER TESEMRTSHE ARGAREEPTV QQAQRRSAVF LSFKSPIPCL PLRWEQQSKL LPTVAGIPAS KVSKWSTDEV SEFIQSLPGC EEHGKVFKDE QIDGEAFLLM TQTDIVKIMS IKLGPALKIF NSILMFKAAE KNSHNEL (400-755 aa encoded by BC060845 )
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt-like protein 3; L(3)mbt-like protein 3; Lethal(3)malignant brain tumor-like protein 3; MBT-1
Gene Aliases: AI481284; KIAA1798; L3MBTL3; MBT-1; MBT1
UniProt ID: (Human) Q96JM7, (Mouse) Q8BLB7
Entrez Gene ID: (Human) 84456, (Mouse) 237339
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support