Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTESASSTSG QEFDVFSVMD WKDGVGTLPG SDLKFRVNEF GALEVITDEN EMENVKKATA TTTWMVPTAQ EVFSEKTGMP FRLKDPVKVE GLQFCENCCQ YGNVDECLSG GNYCSQNCAR HIKDKDQKEE RDVEEDNEEE DPKCSRKKKP KLSLKADNKE DGEERDDEME NKQDVRILRG SQRARRKRRG DSAVLKQGLP PKGKKAWCWA SYLEEEKAVA VPAKLFKEHQ SFPYNKNGFK VGMKLEGVDP EHQSVYCVLT VAEVCGYRIK LHFDGYSDCY DFWVNADALD IHPVGWCEKT GHKLHPPKGY KEEEFNWQTY LKTCKAQAAP KSLFENQNIT VIPSGFRVGM KLEAVDKKNP SFICVATVTD MVDNRFLVHF DNWDESYDYW CEASSPHIHP VGWCKEHRRT LITPPGYPNV KHFSWDKYLE ETNSLPAPAR AFKVKPPHGF QKKMKLEVVD KRNPMFIRVA TVADTDDHRV KVHFDGWNNC YDYWIDADSP DIHPVGWCSK TGHPLQPPLS PLELMEASEH GGCSTPGCKG IGHFKRARHL GPHSAANCPY SEINLNKDRI FPDRLSGEMP PASPSFPRNK RTDANESSSS PEIRDQHADD VKEDFEERTE SEMRTSHEAR GAREEPTVQQ AQRRSAVFLS FKSPIPCLPL RWEQQSKLLP TVAGIPASKV SKWSTDEVSE FIQSLPGCEE HGKVFKDEQI DGEAFLLMTQ TDIVKIMSIK LGPALKIFNS ILMFKAAEKN SHNEL
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-l(3)mbt-like protein 3; l(3)mbt-like protein 3; Lethal(3)malignant brain tumor-like protein 3; MBT-1
Gene Aliases: KIAA1798; L3MBTL3; MBT-1; MBT1
UniProt ID: (Human) Q96JM7
Entrez Gene ID: (Human) 84456
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support