Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Anchoring filament protein which is a component of the basement membrane zone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lad-1; LADA; Ladinin-1; linear IgA bullous dermatosis antigen; Linear IgA disease antigen; linear IgA disease antigen homolog
Gene Aliases: LAD; LAD1; LadA
UniProt ID: (Human) O00515
Entrez Gene ID: (Human) 3898
Molecular Function: structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support