Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • LAMP1 (CD107a) Antibodies

            Invitrogen

            LAMP1 Recombinant Rabbit Monoclonal Antibody (107)

            Advanced Verification
            1 Published Figure
            1 Reference
            View all (101) LAMP1 (CD107a) antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # MA5-29385
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            516.00
            100 µL
             
            In stock
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite LAMP1 Recombinant Rabbit Monoclonal Antibody (107)

            • Antibody Testing Data (11)
            • Published Figures (1)
            • Advanced Verification (1)
            Application
            LAMP1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 13

            LAMP1 Antibody (MA5-29385) in ICC/IF

            Immunofluorescence analysis of LAMP1 (Lysosome-associated membrane glycoprotein 1) was performed using 80% confluent log phase Hep G2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with LAMP1 Recombinant Rabbit Monoclonal Antibody (107) (Product # MA5-29385) at 1:200 in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Rabbit IgG (H+L) Highly Cross-A... View More
            View Product

            LAMP1 Antibody in Immunocytochemistry (ICC/IF)
            LAMP1 Antibody in Immunocytochemistry (ICC/IF)
            LAMP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LAMP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LAMP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LAMP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LAMP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            LAMP1 Antibody in Western Blot (WB)
            LAMP1 Antibody in Western Blot (WB)
            LAMP1 Antibody in Flow Cytometry (Flow)
            LAMP1 Antibody in Immunoprecipitation (IP)
            LAMP1 Antibody in Immunocytochemistry (ICC/IF)
            LAMP1 Antibody

            Product Details

            MA5-29385

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:20-1:100
            View 1 publication 1 publication

            Flow Cytometry (Flow)

            1:20-1:100
            -

            ELISA (ELISA)

            1:5,000-1:10,000
            -

            Immunoprecipitation (IP)

            0.2-1 µL/mg of lysate
            -
            Product Specifications

            Species Reactivity

            Rat

            Published species

            Mouse

            Host/Isotype

            Rabbit / IgG

            Expression System

            HEK293 cells

            Class

            Recombinant Monoclonal

            Type

            Antibody

            Clone

            107

            Immunogen

            Recombinant Human LAMP1 protein (Met1-Met382)
            View immunogen

            Conjugate

            Unconjugated
            • APC 
            • FITC
            • PE 
            • Request custom conjugation

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS

            Contains

            no preservative

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2785256

            Product Specific Information

            This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

            Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

            This antibody has specificity for Human LAMP1/CD107a.

            Target Information

            LAMP1 (CD107a, lysosome-associated membrane protein-1) together with LAMP-2, is a major constituent of lysosomal membrane, 1-2% of total CD107a is found also on the plasma membrane. LAMP1 is a heavily glycosylated membrane protein which contains a putative signal peptide, 18 sites for N-linked glycosylation, a single membrane-spanning segment and a short (11 amino acid) cytosolic tail. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes. LAMP1 is a type 1 integral membrane protein that is transported from trans-Golgi network to endosomes and then lysosomes. Upon cell activation, LAMP1 transfer to the plasma membrane is dependent on a carboyxl-terminal tyrosine based motif (YXXI). Perturbation in the spacing between the tyrosine based motif relative to the membrane abolishes lysosome localization of LAMP1, and this mutant protein then cycles between the plasma membrane and the endosome. Cell surface LAMP1 (and LAMP2) have been shown to promote adhesion of human peripheral blood mononuclear cells (PBMC) to vascular endothelium, therefore, they are possibly involved in the adhesion of PBMC to the site of inflammation. Increased LAMP1 immunoreactivity is observed in neurons and glial cells surrounding senile plaques in Alzheimer’s Disease (AD) cases, and is localized in medullary epithelial cells, single macrophages and lymphocytes in acute thymic involution. LAMP1 is a good marker of mast cell activation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: 120 kDa lysosomal membrane glycoprotein; CD107 antigen-like family member A; CD107a; LAMP-1; LGP-120; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; Lysosome-associated membrane glycoprotein 1; lysosome-associated membrane protein 1

            View more View less

            Gene Aliases: Lamp-1; Lamp1; LGP120

            View more View less

            UniProt ID: (Rat) P14562

            View more View less

            Entrez Gene ID: (Rat) 25328

            View more View less

            Function(s)

            molecular_function enzyme binding protein domain specific binding membrane trafficking regulatory protein

            Process(es)

            autophagy granzyme-mediated apoptotic signaling pathway positive regulation of natural killer cell degranulation positive regulation of natural killer cell mediated cytotoxicity autophagic cell death protein stabilization establishment of protein localization to organelle Golgi to lysosome transport regulation of organelle transport along microtubule
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0