Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SFEDGKPVS LSPLESQAHS PRYTASSQRE RESLEVRMRE VEEENRALRR QLSLAQGRAP SHRRGNHSKT YSMEEGTGDS ENLRAGIVAG NSSECGQQPV VEKCETIHVA IVCAGYNASR DVVTLVKSVL FHRRNPLHFH LIADSIAEQI LATLFQTWM (33-190 aa encoded by BC126404)
The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; like-acetylglucosaminyltransferase; like-glycosyltransferase; myodystrophy; Xylosyl- and glucuronyltransferase LARGE1
Gene Aliases: BPFD#36; enr; fg; froggy; Gyltl1a; KIAA0609; LARGE; LARGE1; Mbp-1; Mbp1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd
UniProt ID: (Human) B0QXZ7, (Mouse) Q9Z1M7
Entrez Gene ID: (Human) 9215, (Mouse) 16795
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support